siRNA Details
| Gene Symbol: DOC2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003585.4 | NP_003576.2 | 8 | 1227 | ||
| XM_011535363.2 | XP_011533665.1 | 7 | 1043 | |||
| GenBank mRNA | BM713588.1 | - | 3 | 236 | ||
| D70830.1 | - | 8 | 1236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DOC2B| Gene Symbol: DOC2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003585.4 | NP_003576.2 | 8 | 1227 | ||
| XM_011535363.2 | XP_011533665.1 | 7 | 1043 | |||
| GenBank mRNA | BM713588.1 | - | 3 | 236 | ||
| D70830.1 | - | 8 | 1236 | |||