siRNA Details
| Gene Symbol: Slc7a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107673.2 | NP_001101143.2 | 11 | 1464 | ||
| XM_006232323.2 | XP_006232385.1 | 10 | 1658 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a11| Gene Symbol: Slc7a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107673.2 | NP_001101143.2 | 11 | 1464 | ||
| XM_006232323.2 | XP_006232385.1 | 10 | 1658 | |||