siRNA Details
| Gene Symbol: Nhlrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761017.1 | XP_008759239.1 | 6 | 960 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nhlrc3| Gene Symbol: Nhlrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761017.1 | XP_008759239.1 | 6 | 960 | ||