siRNA Details
| Gene Symbol: Frem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001245978.1 | NP_001232907.1 | 1 | 4419 | ||
| GenBank mRNA | AB612236.1 | - | 1 | 4419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frem2| Gene Symbol: Frem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001245978.1 | NP_001232907.1 | 1 | 4419 | ||
| GenBank mRNA | AB612236.1 | - | 1 | 4419 | ||