siRNA Details
| Gene Symbol: Plch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191707.1 | NP_001178636.1 | 18 | 2390 | ||
| XM_006232442.2 | XP_006232504.1 | 20 | 4060 | |||
| XM_006232446.2 | XP_006232508.1 | 19 | 3351 | |||
| XM_008761049.1 | XP_008759271.1 | 19 | 3351 | |||
| GenBank mRNA | FQ211591.1 | - | 19 | 2738 | ||

Plch1
Genomic Map