siRNA Details
| Gene Symbol: Nmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107682.2 | NP_001101152.1 | 7 | 709 | ||
| XM_008761090.1 | XP_008759312.1 | 7 | 704 | |||
| GenBank mRNA | BC161998.1 | - | 7 | 709 | 9035992 | |
| FQ215207.1 | - | 5 | 446 | |||

Nmd3
Genomic Map