siRNA Details
| Gene Symbol: Slitrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107683.1 | NP_001101153.1 | 2 | 1869 | ||
| XM_006232490.2 | XP_006232552.1 | 2 | 2139 | |||
| XM_008761091.1 | XP_008759313.1 | 2 | 1611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slitrk3| Gene Symbol: Slitrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107683.1 | NP_001101153.1 | 2 | 1869 | ||
| XM_006232490.2 | XP_006232552.1 | 2 | 2139 | |||
| XM_008761091.1 | XP_008759313.1 | 2 | 1611 | |||