siRNA Details
| Gene Symbol: Golim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191567.1 | NP_001178496.1 | 16, 17 | 2234 | ||
| XM_006232499.2 | XP_006232561.1 | 15, 16 | 2704 | |||
| XM_006232500.1 | XP_006232562.1 | 14, 15 | 2617 | |||
| GenBank mRNA | BC091440.1 | - | 8, 9 | 1188 | ||

Golim4
Genomic Map