siRNA Details
| Gene Symbol: Fnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271167.1 | NP_001258096.1 | 10, 11 | 1217 | ||
| XM_008761099.1 | XP_008759321.1 | 11, 12 | 2702 | |||
| XM_008761100.1 | XP_008759322.1 | 11, 12 | 1334 | |||
| XM_008761101.1 | XP_008759323.1 | 10, 11 | 2612 | |||

Fnip2
Genomic Map