siRNA Details
| Gene Symbol: Fnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271167.1 | NP_001258096.1 | 13 | 1969 | ||
| XM_008761099.1 | XP_008759321.1 | 14 | 3454 | |||
| XM_008761100.1 | XP_008759322.1 | 14 | 2086 | |||
| XM_008761101.1 | XP_008759323.1 | 13 | 3364 | |||

Fnip2
Genomic Map