siRNA Details
| Gene Symbol: Map9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135716.1 | NP_001129188.1 | 12 | 1814 | ||
| XM_006232515.2 | XP_006232577.1 | 12 | 1876 | |||
| XM_006232516.2 | XP_006232578.1 | 11 | 1728 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map9| Gene Symbol: Map9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135716.1 | NP_001129188.1 | 12 | 1814 | ||
| XM_006232515.2 | XP_006232577.1 | 12 | 1876 | |||
| XM_006232516.2 | XP_006232578.1 | 11 | 1728 | |||