siRNA Details
| Gene Symbol: Sfrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100700.1 | NP_001094170.1 | 1 | 453 | ||
| GenBank mRNA | BC161900.1 | - | 1 | 453 | 8367121 | |
More Information
Other Species Targeted
Set Membership: |
|

Sfrp2
Genomic Map