siRNA Details
| Gene Symbol: Sprr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107686.1 | NP_001101156.1 | 1 | 346 | ||
| XM_008761142.1 | XP_008759364.1 | 2 | 396 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprr3| Gene Symbol: Sprr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107686.1 | NP_001101156.1 | 1 | 346 | ||
| XM_008761142.1 | XP_008759364.1 | 2 | 396 | |||