siRNA Details
Gene Symbol: RGD1564386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232801.1 | XP_006232863.1 | 1 | 400 |
Gene Symbol: RGD1564313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749331.1 | XP_003749379.1 | 1 | 487 |
