siRNA Details
| Gene Symbol: Pglyrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191708.1 | NP_001178637.1 | 7 | 854 | ||
| XM_008761143.1 | XP_008759365.1 | 8 | 948 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pglyrp4| Gene Symbol: Pglyrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191708.1 | NP_001178637.1 | 7 | 854 | ||
| XM_008761143.1 | XP_008759365.1 | 8 | 948 | |||