siRNA Details
Gene Symbol: Ilf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047886.1 | NP_001041351.1 | 11, 12 | 1023 | ||
XM_006232624.2 | XP_006232686.1 | 10, 11 | 878 | |||
GenBank mRNA | AY325142.1 | - | 11, 12 | 1023 | ||
BC167105.1 | - | 10, 11 | 778 | 9102039 |
