siRNA Details
Gene Symbol: Ilf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047886.1 | NP_001041351.1 | 4 | 381 | ||
GenBank mRNA | AY325142.1 | - | 4 | 381 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ilf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047886.1 | NP_001041351.1 | 4 | 381 | ||
GenBank mRNA | AY325142.1 | - | 4 | 381 |