siRNA Details
| Gene Symbol: Rhbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183054.1 | NP_898877.1 | 7 | 1121 | ||
| GenBank mRNA | AY129072.1 | - | 7 | 1121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhbg| Gene Symbol: Rhbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183054.1 | NP_898877.1 | 7 | 1121 | ||
| GenBank mRNA | AY129072.1 | - | 7 | 1121 | ||