siRNA Details
| Gene Symbol: Rhbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183054.1 | NP_898877.1 | 6 | 918 | ||
| XM_006232631.2 | XP_006232693.1 | 6 | 1056 | |||
| XM_006232633.2 | XP_006232695.1 | 6 | 1406 | |||
| GenBank mRNA | AY129072.1 | - | 6 | 918 | ||
| BC079365.1 | - | 6 | 900 | 7109321 | ||

Rhbg
Genomic Map