siRNA Details
| Gene Symbol: Tmem79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033896.1 | NP_001029068.1 | 2 | 409 | ||
| XM_006232637.2 | XP_006232699.1 | 2 | 463 | |||
| XM_008761149.1 | XP_008759371.1 | 1 | 928 | |||
| GenBank mRNA | BC101913.1 | - | 2 | 409 | 7936201 | |

Tmem79
Genomic Map