siRNA Details
| Gene Symbol: Sema4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012078.2 | NP_001012078.2 | 10 | 1274 | ||
| XM_006232639.2 | XP_006232701.1 | 10 | 1154 | |||
| XM_006232640.2 | XP_006232702.1 | 10 | 1125 | |||
| XM_006232641.2 | XP_006232703.1 | 10 | 1354 | |||
| XM_008761150.1 | XP_008759372.1 | 10 | 1848 | |||
| GenBank mRNA | BC081943.1 | - | 10 | 1218 | 7121310 | |

Sema4a
Genomic Map