siRNA Details
| Gene Symbol: Ash1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107689.1 | NP_001101159.1 | 3 | 3175 | ||
| XM_008761153.1 | XP_008759375.1 | 3 | 3150 | |||
| XM_008761154.1 | XP_008759376.1 | 2 | 3018 | |||
| XM_008761155.1 | XP_008759377.1 | 3 | 3302 | |||
| XM_008761156.1 | XP_008759378.1 | 4 | 3437 | |||
| XM_008761157.1 | XP_008759379.1 | 4 | 3323 | |||
| XM_008761158.1 | XP_008759380.1 | 3 | 3219 | |||
| XM_008761159.1 | XP_008759381.1 | 3 | 3067 | |||
| XM_008761160.1 | XP_008759382.1 | 2 | 3018 | |||

Ash1l
Genomic Map