siRNA Details
| Gene Symbol: Tdrkh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014038.2 | NP_001014060.2 | 12 | 1690 | ||
| XM_006232870.2 | XP_006232932.1 | 12 | 1693 | |||
| XM_006232871.2 | XP_006232933.1 | 12 | 3649 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdrkh| Gene Symbol: Tdrkh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014038.2 | NP_001014060.2 | 12 | 1690 | ||
| XM_006232870.2 | XP_006232932.1 | 12 | 1693 | |||
| XM_006232871.2 | XP_006232933.1 | 12 | 3649 | |||