siRNA Details
| Gene Symbol: Vps72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107695.1 | NP_001101165.1 | 5 | 644 | ||
| GenBank mRNA | BC161999.1 | - | 5 | 632 | 9036078 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps72| Gene Symbol: Vps72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107695.1 | NP_001101165.1 | 5 | 644 | ||
| GenBank mRNA | BC161999.1 | - | 5 | 632 | 9036078 | |