siRNA Details
| Gene Symbol: Prune | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007697.1 | NP_001007698.1 | 3, 4 | 583 | ||
| XM_006232885.2 | XP_006232947.1 | 3, 4 | 595 | |||
| GenBank mRNA | BC079054.1 | - | 3, 4 | 583 | 7115417 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prune| Gene Symbol: Prune | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007697.1 | NP_001007698.1 | 3, 4 | 583 | ||
| XM_006232885.2 | XP_006232947.1 | 3, 4 | 595 | |||
| GenBank mRNA | BC079054.1 | - | 3, 4 | 583 | 7115417 | |