siRNA Details
| Gene Symbol: Otud7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107697.1 | NP_001101167.1 | 7 | 1173 | ||
| XM_006232957.2 | XP_006233019.1 | 7 | 942 | |||
| XM_006232958.2 | XP_006233020.1 | 6 | 750 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otud7b| Gene Symbol: Otud7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107697.1 | NP_001101167.1 | 7 | 1173 | ||
| XM_006232957.2 | XP_006233019.1 | 7 | 942 | |||
| XM_006232958.2 | XP_006233020.1 | 6 | 750 | |||