siRNA Details
| Gene Symbol: Hist2h3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107698.1 | NP_001101168.1 | 1 | 315 | ||
| XM_008761309.1 | XP_008759531.1 | 1 | 372 | |||
| Gene Symbol: LOC690126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761346.1 | XP_008759568.1 | 1 | 367 | ||

Hist2h3c2
Genomic Map