siRNA Details
| Gene Symbol: Wdr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107705.1 | NP_001101175.1 | 13 | 1543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr3| Gene Symbol: Wdr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107705.1 | NP_001101175.1 | 13 | 1543 | ||