siRNA Details
| Gene Symbol: Fam46c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014041.1 | NP_001014063.1 | 2 | 633 | ||
| XM_006233038.2 | XP_006233100.1 | 2 | 451 | |||
| XM_006233039.2 | XP_006233101.1 | 2 | 555 | |||
| XM_008761361.1 | XP_008759583.1 | 2 | 423 | |||
| GenBank mRNA | BC083570.1 | - | 2 | 633 | 7111947 | |

Fam46c
Genomic Map