siRNA Details
| Gene Symbol: Slc22a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107707.1 | NP_001101177.1 | 8 | 1338 | ||
| XM_006233050.2 | XP_006233112.1 | 7 | 3732 | |||
| GenBank mRNA | FQ211559.1 | - | 4 | 919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a15| Gene Symbol: Slc22a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107707.1 | NP_001101177.1 | 8 | 1338 | ||
| XM_006233050.2 | XP_006233112.1 | 7 | 3732 | |||
| GenBank mRNA | FQ211559.1 | - | 4 | 919 | ||