siRNA Details
| Gene Symbol: Olfml3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107708.1 | NP_001101178.1 | 2 | 450 | ||
| XM_006233077.1 | XP_006233139.1 | 2 | 435 | |||
| GenBank mRNA | BC158838.1 | - | 2 | 389 | ||
| FQ228335.1 | - | 2 | 402 | |||

Olfml3
Genomic Map