siRNA Details
| Gene Symbol: Dclre1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025687.1 | NP_001020858.1 | 3 | 444 | ||
| GenBank mRNA | BC098939.1 | - | 3 | 444 | 7455348 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dclre1b| Gene Symbol: Dclre1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025687.1 | NP_001020858.1 | 3 | 444 | ||
| GenBank mRNA | BC098939.1 | - | 3 | 444 | 7455348 | |