siRNA Details
| Gene Symbol: Ap4b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107709.1 | NP_001101179.1 | 9 | 2046 | ||
| XM_006233081.2 | XP_006233143.1 | 10 | 1913 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap4b1| Gene Symbol: Ap4b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107709.1 | NP_001101179.1 | 9 | 2046 | ||
| XM_006233081.2 | XP_006233143.1 | 10 | 1913 | |||