siRNA Details
| Gene Symbol: Cttnbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107712.2 | NP_001101182.2 | 3 | 221 | ||
| XM_006233087.1 | XP_006233149.1 | 2 | 198 | |||
| GenBank mRNA | CB724536.1 | - | 3 | 239 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cttnbp2nl| Gene Symbol: Cttnbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107712.2 | NP_001101182.2 | 3 | 221 | ||
| XM_006233087.1 | XP_006233149.1 | 2 | 198 | |||
| GenBank mRNA | CB724536.1 | - | 3 | 239 | ||