siRNA Details
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008771.3 | NP_001008771.2 | 10 | 1073 | ||
| GenBank mRNA | AY321348.1 | - | 11 | 1034 | ||
| BC097367.1 | - | 10 | 1024 | 7443204 | ||
| FQ213175.1 | - | 10 | 1168 | |||
| FQ226328.1 | - | 10 | 1068 | |||
| Gene Symbol: LOC100911453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749373.3 | XP_003749421.1 | 10 | 1179 | ||

Wdr77
Genomic Map