siRNA Details
| Gene Symbol: Wdr47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100702.1 | NP_001094172.1 | 13 | 2601 | ||
| XM_006233155.2 | XP_006233217.1 | 13 | 2755 | |||
| XM_006233157.2 | XP_006233219.1 | 13 | 2731 | |||
| XM_008761402.1 | XP_008759624.1 | 13 | 2673 | |||
| XM_008761403.1 | XP_008759625.1 | 13 | 2752 | |||
| GenBank mRNA | BC091374.1 | - | 4 | 618 | ||

Wdr47
Genomic Map