siRNA Details
| Gene Symbol: Slc25a24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127544.1 | NP_001121016.1 | 8 | 1142 | ||
| XM_008761404.1 | XP_008759626.1 | 7 | 1170 | |||
| GenBank mRNA | BC162022.1 | - | 8 | 1121 | 9098607 | |
| FQ211892.1 | - | 7 | 1102 | |||
| FQ221591.1 | - | 8 | 1222 | |||

Slc25a24
Genomic Map