siRNA Details
| Gene Symbol: Slc30a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191715.1 | NP_001178644.1 | 8 | 941 | ||
| GenBank mRNA | BC091417.1 | - | 4 | 299 | ||
| BC107441.1 | - | 4 | 300 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a7| Gene Symbol: Slc30a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191715.1 | NP_001178644.1 | 8 | 941 | ||
| GenBank mRNA | BC091417.1 | - | 4 | 299 | ||
| BC107441.1 | - | 4 | 300 | |||