siRNA Details
| Gene Symbol: Extl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100704.1 | NP_001094174.1 | 5 | 745 | ||
| XM_006233215.2 | XP_006233277.1 | 5 | 1290 | |||
| XM_006233216.2 | XP_006233278.1 | 5 | 1031 | |||
| XM_006233217.2 | XP_006233279.1 | 4 | 822 | |||
| GenBank mRNA | BC162014.1 | - | 5 | 745 | 9091843 | |

Extl2
Genomic Map