siRNA Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012082.1 | NP_001012082.1 | 8 | 1076 | ||
| XM_008761461.1 | XP_008759683.1 | 8 | 1344 | |||
| GenBank mRNA | BC079371.1 | - | 8 | 1076 | 7109915 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35a3| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012082.1 | NP_001012082.1 | 8 | 1076 | ||
| XM_008761461.1 | XP_008759683.1 | 8 | 1344 | |||
| GenBank mRNA | BC079371.1 | - | 8 | 1076 | 7109915 | |