siRNA Details
| Gene Symbol: Ptbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005555.1 | NP_001005555.1 | 8 | 902 | ||
| XM_006233236.2 | XP_006233298.1 | 8 | 1436 | |||
| XM_006233237.2 | XP_006233299.1 | 8 | 1435 | |||
| XM_006233238.2 | XP_006233300.1 | 8 | 1435 | |||
| GenBank mRNA | AJ010585.1 | - | 8 | 871 | ||
| BC082076.1 | - | 8 | 902 | 7133221 | ||
| FQ212405.1 | - | 8 | 916 | |||

Ptbp2
Genomic Map