siRNA Details
Gene Symbol: Arhgap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009405.1 | NP_001009405.1 | 20 | 2392 | ||
XM_006233239.2 | XP_006233301.1 | 19 | 4192 | |||
XM_006233240.2 | XP_006233302.1 | 19 | 2381 | |||
XM_006233241.2 | XP_006233303.1 | 19 | 2631 | |||
GenBank mRNA | BC087167.1 | - | 20 | 2392 | 7135494 |
