siRNA Details
| Gene Symbol: Arhgap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009405.1 | NP_001009405.1 | 18 | 2003 | ||
| XM_006233239.2 | XP_006233301.1 | 17 | 3803 | |||
| XM_006233240.2 | XP_006233302.1 | 17 | 1992 | |||
| XM_006233241.2 | XP_006233303.1 | 17 | 2242 | |||
| GenBank mRNA | BC087167.1 | - | 18 | 2003 | 7135494 | |

Arhgap29
Genomic Map