siRNA Details
| Gene Symbol: Arhgap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009405.1 | NP_001009405.1 | 21 | 2597 | ||
| XM_006233239.2 | XP_006233301.1 | 20 | 4397 | |||
| XM_006233240.2 | XP_006233302.1 | 20 | 2586 | |||
| XM_006233241.2 | XP_006233303.1 | 20 | 2836 | |||
| GenBank mRNA | BC087167.1 | - | 21 | 2597 | 7135494 | |

Arhgap29
Genomic Map