siRNA Details
| Gene Symbol: Abca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107721.1 | NP_001101191.1 | 24 | 3598 | ||
| XM_006233242.2 | XP_006233304.1 | 24 | 3777 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca4| Gene Symbol: Abca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107721.1 | NP_001101191.1 | 24 | 3598 | ||
| XM_006233242.2 | XP_006233304.1 | 24 | 3777 | |||