siRNA Details
| Gene Symbol: Bcar3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107722.1 | NP_001101192.1 | 11 | 2632 | ||
| XM_006233270.2 | XP_006233332.1 | 9 | 3029 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcar3| Gene Symbol: Bcar3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107722.1 | NP_001101192.1 | 11 | 2632 | ||
| XM_006233270.2 | XP_006233332.1 | 9 | 3029 | |||