siRNA Details
| Gene Symbol: Fnbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039609.1 | NP_001034698.1 | 322 | |||
| GenBank mRNA | AB250295.1 | - | 322 | |||
| AB250296.1 | - | 322 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fnbp1l| Gene Symbol: Fnbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039609.1 | NP_001034698.1 | 322 | |||
| GenBank mRNA | AB250295.1 | - | 322 | |||
| AB250296.1 | - | 322 | ||||