siRNA Details
| Gene Symbol: Tifa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014044.1 | NP_001014066.1 | 3 | 383 | ||
| XM_006233289.2 | XP_006233351.1 | 3 | 573 | |||
| XM_006233290.2 | XP_006233352.1 | 3 | 550 | |||
| XM_008761463.1 | XP_008759685.1 | 3 | 722 | |||
| XM_008761464.1 | XP_008759686.1 | 3 | 482 | |||
| GenBank mRNA | AY325204.1 | - | 826 | |||
| BC083765.1 | - | 3 | 383 | 7193165 | ||

Tifa
Genomic Map