siRNA Details
| Gene Symbol: Adh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012084.1 | NP_001012084.1 | 3, 4 | 307 | ||
| XM_006233372.2 | XP_006233434.1 | 3, 4 | 1101 | |||
| XM_006233374.1 | XP_006233436.1 | 4, 5 | 379 | |||
| XM_006233375.2 | XP_006233437.1 | 5, 6 | 590 | |||
| XM_006233377.2 | XP_006233439.1 | 3, 4 | 549 | |||
| XM_006233378.2 | XP_006233440.1 | 4, 5 | 459 | |||
| XM_006233379.2 | XP_006233441.1 | 4, 5 | 456 | |||
| XM_008761506.1 | XP_008759728.1 | 2, 3 | 421 | |||
| XM_008761507.1 | XP_008759729.1 | 4, 5 | 2653 | |||
| XM_008761508.1 | XP_008759730.1 | 5, 6 | 2513 | |||
| XM_008761509.1 | XP_008759731.1 | 5, 6 | 1005 | |||
| XM_008761510.1 | XP_008759732.1 | 5, 6 | 2781 | |||
| GenBank mRNA | BC083792.1 | - | 3, 4 | 307 | 7106499 | |

Adh6
Genomic Map