siRNA Details
| Gene Symbol: Rap1gds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107728.1 | NP_001101198.1 | 4 | 483 | ||
| XM_006233395.2 | XP_006233457.1 | 2 | 2543 | |||
| XM_008761511.1 | XP_008759733.1 | 2 | 2547 | |||
| XM_008761512.1 | XP_008759734.1 | 3 | 262 | |||

Rap1gds1
Genomic Map